Orders LinkContactUsLinkComponent
SYPRO® Ruby protein blot stain
SYPRO® Ruby protein blot stain
SYPRO® Ruby Protein Blot Stain, 200 mL
Katalog # LONZ50565
Leverandør:   LONZA
SYPRO® Ruby protein blot stain
Katalog # LONZ50565
Leverandør:  LONZA
CAS nummer:  
SYPRO® Ruby protein blot stain
SYPRO® Ruby Protein Blot Stain, 200 mL
CAS nummer:  
Price
3.820,00 kr
Log ind for tilgængelighed
1 * 200 mL
Product Details & Documents
SYPRO™ Ruby is 60-times more sensitive than reversible protein blot stains like Ponceau S, and 30-times more sensitive than Amido Black or Coomassie® brilliant blue stains.
  • Visualize proteins prior to transfer - does not interfere with protein activity
  • Versatile - Excellent for staining native proteins
  • Safe - No acids or organic solvents necessary
  • Sensitive - Detects as little as 4-8 ng protein per band
Specifikationer
Specifikationer
  • Pakn:
    200 mL
  • Beskrivelse:
    SYPRO® Ruby Protein Blot Stain