Micro particles based on polystyrene, analytical standard, Supelco®
Supplier: Merck
About this item
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
7 Options Available Below
Product Details & Documents
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 74964-5ML-F
- 74964-10ML-F
- 74491-5ML-F
- 74491-10ML-F
- 79807-5ML-F
- 84135-5ML-F
- 84135-10ML-F
- M-Clarity quality level:
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
Specifications
Product Family Options
Product Information
- PartikelstørrelseBeskrivelseNøjagtighedStandardPaknAvailabilityPrice
- 15 µmAqueous suspension, 10% solidsstd dev <0,2 µmAnalytical standard5 mLLog in for availability1 * 5 mL2.370,00 krSpecifications:
More
Specifications
Cat. No.74964-5ML-FM-Clarity quality levelMQ100 - 15 µmAqueous suspension, 10% solidsstd dev <0,2 µmAnalytical standard10 mLLog in for availability1 * 10 mL3.510,00 krSpecifications:
More
Specifications
Cat. No.74964-10ML-FM-Clarity quality levelMQ100 - 20 µmAqueous suspension, 10% solidsstd dev <0,3 µmAnalytical standard5 mLLog in for availability1 * 5 mL2.270,00 krSpecifications:
More
Specifications
Cat. No.74491-5ML-FM-Clarity quality levelMQ100 - 20 µmAqueous suspension, 10% solidsstd dev <0,3 µmAnalytical standard10 mLLog in for availability1 * 10 mL3.790,00 krSpecifications:
More
Specifications
Cat. No.74491-10ML-FM-Clarity quality levelMQ100 - 25 µmAqueous suspension, 10% solidsstd dev <0,4 µmAnalytical standard5 mLLog in for availability1 * 5 mL2.270,00 krSpecifications:
More
Specifications
Cat. No.79807-5ML-FM-Clarity quality levelMQ100 - 30 µmAqueous suspension, 10% solidsstd dev <0,4 µmAnalytical standard5 mLLog in for availability1 * 5 mL2.430,00 krSpecifications:
More
Specifications
Cat. No.84135-5ML-FM-Clarity quality levelMQ100 - 30 µmAqueous suspension, 10% solidsstd dev <0,4 µmAnalytical standard10 mLLog in for availability1 * 10 mL3.790,00 krSpecifications:
More
Specifications
Cat. No.84135-10ML-FM-Clarity quality levelMQ100