Chloride standard for IC, TraceCERT®, Supelco®
Catalog # 06740-100ML
Supplier: Merck
CAS Number:
Specifications
- Koncentration:10000 mg/l
- Pakn:100 mL
- Beskrivelse:Chloride in water
Specifications
About this item
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.