Mop heads, dry
Mop head, NovaPoly, Novalite, 356 mm
MICRMZDM62-14
Mops absorb detergent quickly and may be used on floors, walls, and ceilings without a bucket or wringer.
- May be used with disinfectants
Material is designed for applications in which excess moisture is a concern. A special fabric strip allows for fast-drying action. Mop heads are sturdy enough to be laundered and reused. Latex/rayon blend mop head provides absorbency like that of PVA.
- Beskrivelse:Mop head, NovaPoly, Novalite
- Længde:356 mm
- Seq:04