Supelco® Organic Reference Standard, Oleanolic Acid
Oleanolic acid, Analytical standard, Neat, 10 mg
Leverandør: Merck
Supelco® Organic Reference Standard, Oleanolic Acid
Katalog # 42515-10MG
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Emballering: Bottomless glass bottle. Contents are inside inserted fused cone.
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:10 mg
- Standard:Analytical standard
- Beskrivelse:Oleanolic acid
- Form:Neat
- Cat. No.:42515-10MG
- M-Clarity quality level:MQ100
- CAS-nr:508-02-1
- Molekylvægt:456,70
- MDL No.:MFCD00064914
- Opbevaringstemperatur:2...8 °C
- Flammepunkt:Not applicable
- Opbevaring betingelser:2...8 °C
- Melting point:>300 °C (lit.)
- Shipping conditions:WICE
- Formel:C₃₀H₄₈O₃



