Supelco® Organic Reference Standard, Khellin
Khellin, Analytical standard, 100 mg
Katalog # 00899-100MG
Leverandør: Merck
Gå til:
Product Details & Documents
- GHS signal: GHS06
Caution: P261 - P280 - P301 + P310 - P302 + P352 + P312 - P304 + P340 + P311 - P305 + P351 + P338
Specifikationer
Specifikationer
- Pakn:100 mg
- Standard:Analytical standard
- Beskrivelse:Khellin
- MDL No.:MFCD00005007
- Synonym(s):4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one
- Formel:C₁₄H₁₂O₅
- Cat. No.:00899-100MG
- M-Clarity quality level:MQ100
- CAS-nr:82-02-0
- Molekylvægt:260,24
- Kogepunkt:180...200 °C/0,05 mm Hg (lit.)
- Fareklassebeskrivelse:H301 + H311 + H331 - H315 - H319 - H335
- Flammepunkt:Not applicable
- Opbevaring betingelser:2...30 °C
- Melting point:150...154 °C (lit.)
- Shipping conditions:STD
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



