Orders LinkContactUsLinkComponent
Cylinder brushes
Cylinder brushes
Leverandør:   MICRONOVA
Cylinder brushes
Leverandør:  MICRONOVA
Certifikater
Cylinder brushes
2 tilgængelige muligheder nedenfor
Product Details & Documents
These cylinder brushes with brush heads feature ACME thread for attachment to STA series handles.
  • With optional fabric covers
  • Polyester bristles
  • BPA-free
Base resins used to produce blocks and bristles are made from FDA approved materials.
Polyester cover is compatible with strong disinfectants and autoclavable.
Product Family Options
Produktinformation
  • Børstemateriale
    Børstehoved Ø
    Handle material
    Tilgængelighed
    Price
  • MICRBRWH-03
    Cylinder brush
    Cylinder brush
    Polyester
    127 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 961,00 kr
    961,00 kr
    1 * 1 Items
       961,00 kr
  • MICRBRWH-02
    Cylinder brush
    Cylinder brush
    Polyester
    178 mm
    Plast
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 961,00 kr
    961,00 kr
    1 * 1 Items
       961,00 kr
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Polymesh cover, for use with 178 mm brush, 100% polyester covered in polyester abrasive mesh
    Polymesh cover, for use with 178 mm brush, 100% polyester covered in polyester abrasive mesh
    MICRBRSC68-02
    Log ind for tilgængelighed
    1 * 12 Items - 3.670,00 kr