Orders LinkContactUsLinkComponent
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Osteo-Bed Bone Embedding Kit, Sigma-Aldrich®, 1 KIT
Katalog # EM0200-1KT
Leverandør:   SIGMA ALDRICH MICROSCOPY
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Katalog # EM0200-1KT
CAS nummer:  
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Osteo-Bed Bone Embedding Kit, Sigma-Aldrich®, 1 KIT
CAS nummer:  
Price
2.850,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Suitable for use with large and small mineralized (undecalcified) bone sections. Yields clear, hard blocks for cutting sections. Not water-soluble.
  • Quality Level 200
Kit contains:
Osteo-Bed Bone Embedding Resin A (O8514) 900 ml
Osteo-Bed Bone Embedding Catalyst (O8764) 2 x 12 g
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Osteo-Bed Bone Embedding Kit, Sigma-Aldrich®