Embedding kit JB-4, Sigma-Aldrich®
Katalog # EM0100-1KT
Leverandør: SIGMA ALDRICH MICROSCOPY
CAS nummer:
Specifikationer
- Pakn:1 KIT
- Beskrivelse:Embedding kit JB-4, Sigma-Aldrich®
- Environmentally Preferable:
Specifikationer
Om denne vare
JB-4 is a water-soluble, GMA based, plastic resin kit intended for use in the preparation of embedded samples for high resolution light microscopy. It is a technique now widely used for research and clinical diagnosis. JB-4 yields semi-thin sections (0.5μ-2μ) with excellent morphological preservation.
- Quality Level 200
JB-4 kit contains:
JB-4 Embedding Solution A (J4954) 800ml
JB-4 Embedding Solution B (J0330) 30 ml
Benzoyl Peroxide (J0455) 12 g