Safety spectacles, 553 Zeronoise
Anti-UV, anti-scratch, anti-fog, Clear PC
Katalog # UNVT553Z.34.00.00
Leverandør: UNIVET
Gå til:
Product Details & Documents
The 553Z model spectacle is equipped with patented temples specially designed to facilitate the combined use of the two protective devices.
- Wraparound model
- Optimal lateral protection
- Light and handy
- Polycarbonate material
- In/out frame and lens
The shorter terminals and the reduced sections cancel the interference with cushions avoiding the earmuff loses part of its attenuation capacity.
The flexible and shaped temples give the operator comfort and allow to wear the glass without having to remove the earmuffs.
Weight: 19 g
Specifikationer
- Model:Anti-UV, anti-scratch, anti-fog
- Linser:Clear PC