Supelco® Organic Reference Standard, (±)-Geosmin solution
(±)-Geosmin solution, 100 µg/ml in methanol, Certified reference material, 1
Katalog # 4M7522-U
Leverandør: Merck
Supelco® Organic Reference Standard, (±)-Geosmin solution
Katalog # 4M7522-U
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
Advarsel: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311
Specifikationer
Specifikationer
- Koncentration:100 µg/ml in methanol
- Pakn:1
- Standard:Certified reference material
- Beskrivelse:(±)-Geosmin solution
- Cat. No.:4M7522-U
- M-Clarity quality level:MQ100
- CAS-nr:16423-19-1
- Molekylvægt:182,30
- Fareklassebeskrivelse:H225 - H301 + H311 + H331 - H370
- Opbevaring betingelser:2...30 °C
- Shipping conditions:STD



