Multi-Purpose Trolleys
Leverandør: MULTIROIR
Om denne vare
These multi-use trolleys have been designed for intensive use. They are made of an aluminium frame with heavy duty polypropylene trays.
- Grey or black
- 2 or 3 tray versions
- Unequalled load capacity: 80 kg per tray and 250 kg per trolley
- 4 ball bearing wheels diam. 100 mm, 2 with brakes
The trolleys are light, easy to handle and very robust.
6 tilgængelige muligheder nedenfor
Product Details & Documents
These multi-use trolleys have been designed for intensive use. They are made of an aluminium frame with heavy duty polypropylene trays.
- Grey or black
- 2 or 3 tray versions
- Unequalled load capacity: 80 kg per tray and 250 kg per trolley
- 4 ball bearing wheels diam. 100 mm, 2 with brakes
The trolleys are light, easy to handle and very robust.
Specifications for Product Family
Specifikationer
- Catalog No:
- 216-2232
- 216-2231
- 216-2234
- 216-2236
- 216-2235
- 216-2238
- Model:
- 65005HS808E3
- 65007HS808E
- 65006HS808A3
- 65001HS808LE3
- 65003HS808LE
- 65002HS808LA3
- Materiale:
- PP
- PP
- PP
- PP
- PP
- PP
- Tray dimensions:
- 615W×450D mm (ext.)
607W×435D mm (int.) - 680W×450D mm (ext.)
670W×435D mm (int.) - 615W×450D mm (ext.)
607W×435D mm (int.) - 920W×520D mm (ext.)
910W×504D mm (int.) - 920W×520D mm (ext.)
910W×504D mm (int.) - 920W×520D mm (ext.)
910W×504D mm (int.)
- Højde:
- 553 mm (height between two trays)
- 553 mm (height between two trays)
- 275 mm (height between two trays)
- 553 mm (height between two trays)
- 553 mm (height between two trays)
- 275 mm (height between two trays)
Specifikationer
Product Family Options
Produktinformation
- B×D×HHylderFarveTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.216-2232Model65005HS808E3MaterialePPTray dimensions615W×450D mm (ext.)
607W×435D mm (int.)Højde553 mm (height between two trays) - Specifikationer:
More
Specifications
Cat. No.216-2231Model65007HS808EMaterialePPTray dimensions680W×450D mm (ext.)
670W×435D mm (int.)Højde553 mm (height between two trays) - Specifikationer:
More
Specifications
Cat. No.216-2234Model65006HS808A3MaterialePPTray dimensions615W×450D mm (ext.)
607W×435D mm (int.)Højde275 mm (height between two trays) - Specifikationer:
More
Specifications
Cat. No.216-2236Model65001HS808LE3MaterialePPTray dimensions920W×520D mm (ext.)
910W×504D mm (int.)Højde553 mm (height between two trays) - Specifikationer:
More
Specifications
Cat. No.216-2235Model65003HS808LEMaterialePPTray dimensions920W×520D mm (ext.)
910W×504D mm (int.)Højde553 mm (height between two trays) - Specifikationer:
More
Specifications
Cat. No.216-2238Model65002HS808LA3MaterialePPTray dimensions920W×520D mm (ext.)
910W×504D mm (int.)Højde275 mm (height between two trays)