Orders LinkContactUsLinkComponent
Neddle holders, Aesculap®
Supplier:   Aesculap
Neddle holders, Aesculap®
Supplier:  Aesculap
Certificates
Neddle holders, Aesculap®
27 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Beskrivelse
    Model
    Længde
    Availability
    Price
  • Needle holder, Converse
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Derf
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Doyen
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Webster
    Smooth jaw 
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 816,00 kr
    816,00 kr
    1 * 1 Items
       816,00 kr
  • Needle holder, Halsey
    Smooth jaw
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Halsey
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Kilner
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Baumgartner
    145 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder with groove, Hegar
    Heavy pattern 
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Crile-Murray
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Crile-Wood
    Fine pattern 
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Hegar-Mayo
    Medium fine pattern 
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Lawrence
    Delicate jaw
    152 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Crile-Wood
    Solid pattern 
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Mayo-Hegar
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder with groove, Hegar
    Heavy pattern 
    175 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Adson
    Fenestrated
    175 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Mayo-Hegar
    Medium fine pattern 
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 514,00 kr
    514,00 kr
    1 * 1 Items
       514,00 kr
  • Needle holder, Mayo-Hegar
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Heaney
    Curved 
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Mayo-Hegar
    Medium fine pattern 
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder with groove, Hegar
    Heavy pattern 
    205 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder without groove, Hegar
    Heavy pattern 
    205 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Jameson
    Straight, slender 
    230 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Masson-Luethy
    Bayonet shaped 
    255 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Masson
    265 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Needle holder, Wangensteen
    275 mm