Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Chromium
VISOCOLOR® Colorimetric Test, Chromium
Leverandør:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Chromium
Leverandør:  MACHEREY-NAGEL
Certifikater
VISOCOLOR® Colorimetric Test, Chromium
2 tilgængelige muligheder nedenfor
Product Details & Documents
Reagents for the determination of chromium in water samples.
  • Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Remark: Measuring range on PF-12Plus. Range on other photometers can be different.
Hazardous material: Yes
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • MANA931020
  • MANA931220
  • Model:
  • MANA931020
  • MANA931220
  • Platform:
  • Colorimetric test kits
  • Colorimetric test kits
  • Opbevaringstemperatur:
  • 15...25 °C
  • 15...25 °C
  • Holdbarhed (måneder):
  • 18
  • 18
  • Vægt:
  • 239,4 g (Including packaging)
  • 149,5 g (Including packaging)
Specifikationer
Product Family Options
Test kits
Produktinformation
  • Indhold
    Område
    Kemisk metode
    Til enhed
    Dimensioner
    Test pr. sæt
    Pakn
    Tilgængelighed
    Price
  • MANA931020
    VISOCOLOR® ECO Chromium(VI), test no. 5-20
    VISOCOLOR® ECO Chromium(VI), test no. 5-20
    Reagents for 140 tests, colour chart and accessories in a box
    0,02 • 0,05 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L; 0,02 - 0,50 mg/L
    Carbazide
    PF-12Plus; PF-3 Fish; Advance; Visual
    168×112×40 mm (Including packaging)
    140
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 666,00 kr
    666,00 kr
    1 * 1 Each
       666,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA931020
    Model
    MANA931020
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15...25 °C
    Holdbarhed (måneder)
    18
    Vægt
    239,4 g (Including packaging)
Refill packs
Produktinformation
  • Indhold
    Område
    Kemisk metode
    Til enhed
    Dimensioner
    Test pr. sæt
    Pakn
    Tilgængelighed
    Price
  • MANA931220
    VISOCOLOR® ECO Chromium(VI), refill pack, test no. 5-20
    VISOCOLOR® ECO Chromium(VI), refill pack, test no. 5-20
    Reagents for 140 tests in a box
    0,02 • 0,05 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L; 0,02 - 0,50 mg/L
    Carbazide
    PF-12Plus; Advance; Visual
    170×115×70 mm (Including packaging)
    140
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 476,00 kr
    476,00 kr
    1 * 1 Each
       476,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA931220
    Model
    MANA931220
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15...25 °C
    Holdbarhed (måneder)
    18
    Vægt
    149,5 g (Including packaging)