Capsule filters, Polycap™ TF, Whatman™
Om denne vare
These Polycap™ TF capsule filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a PP housing for use with organic solvents and chemically aggressive solutions.
- PTFE filter membrane is resistant to most solvents
- 1,0 µm pore size capsules used for extended life and filtration of highly contaminated solutions
- Autoclavable capsule filtration device if sterility is required
Filtration of organic solvents and aggressive solutions requires a durable membrane for filtration without sample contamination.
Autoclavable Polycap™ TF capsule filters feature PTFE membranes with high chemical resistance. Common applications include venting, in-line filtration to sterilise and remove particles from air/gas/liquid streams, filtration to ensure clean inert gases in vial filling lines, and isolation of exhaust from fermenters.
Polycap™ TF filters support use in the electronics, pharmaceutical, and biopharmaceutical industries, because they are photoresistant and can be used to vent and perform in-line filtration for production systems in biological environments.
: Manufactured according to ISO9001:2008 QMS standards
24 tilgængelige muligheder nedenfor
Product Details & Documents
These Polycap™ TF capsule filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a PP housing for use with organic solvents and chemically aggressive solutions.
- PTFE filter membrane is resistant to most solvents
- 1,0 µm pore size capsules used for extended life and filtration of highly contaminated solutions
- Autoclavable capsule filtration device if sterility is required
Filtration of organic solvents and aggressive solutions requires a durable membrane for filtration without sample contamination.
Autoclavable Polycap™ TF capsule filters feature PTFE membranes with high chemical resistance. Common applications include venting, in-line filtration to sterilise and remove particles from air/gas/liquid streams, filtration to ensure clean inert gases in vial filling lines, and isolation of exhaust from fermenters.
Polycap™ TF filters support use in the electronics, pharmaceutical, and biopharmaceutical industries, because they are photoresistant and can be used to vent and perform in-line filtration for production systems in biological environments.
: Manufactured according to ISO9001:2008 QMS standards
Specifications for Product Family
Specifikationer
- Catalog No:
- 516-0911
- 516-4450
- 515-0091
- 515-0152
- 516-0955
- 516-4451
- 516-0956
- 516-0442
- 515-0092
- 516-0962
- 515-0093
- 516-0913
- 516-0963
- 516-4453
- 515-0094
- 516-0917
- 516-0964
- 515-0095
- 515-0096
- 516-0973
- 516-0974
- 516-0975
- 516-0976
- 516-0977
- Model:
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 36, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE, sterile
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 75, PTFE
- Polycap™ TF 150, PTFE
- Polycap™ TF 150, PTFE
- Polycap™ TF 150, PTFE
- Polycap™ TF 150, PTFE
- Polycap™ TF 150, PTFE
- Filtreringsareal:
- 500 cm²
- 500 cm²
- 500 cm²
- 500 cm²
- 500 cm²
- 500 cm²
- 500 cm²
- 500 cm²
- 500 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 1000 cm²
- 2000 cm²
- 2000 cm²
- 2000 cm²
- 2000 cm²
- 2000 cm²
- Maks. tryk:
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
Specifikationer
Product Family Options
Produktinformation
- ModelPorestørrelseFiltreringsområdeTilslutningTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.516-0911ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Polycap™ TF 36, PTFE0,2 µmMNPT/Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items1.110,00 krSpecifikationer:
More
Specifications
Cat. No.516-4450ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Polycap™ TF 36, PTFE0,2 µm500 cm²Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items967,00 krSpecifikationer:
More
Specifications
Cat. No.515-0091ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Polycap™ TF 36, PTFE0,2 µm500 cm²Stepped barb for 10 - 12 mm tubingLog ind for tilgængelighed1 * 1 Items908,00 krSpecifikationer:
More
Specifications
Cat. No.515-0152ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0955ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Polycap™ TF 36, PTFE0,45 µmMNPT/Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items1.070,00 krSpecifikationer:
More
Specifications
Cat. No.516-4451ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0956ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Polycap™ TF 36, PTFE1,0 µmStepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 5 Items2.900,00 krSpecifikationer:
More
Specifications
Cat. No.516-0442ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Polycap™ TF 36, PTFE1,0 µm500 cm²Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items986,00 krSpecifikationer:
More
Specifications
Cat. No.515-0092ModelPolycap™ TF 36, PTFEFiltreringsareal500 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0962ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Polycap™ TF 75, PTFE0,1 µm1000 cm²Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items1.680,00 krSpecifikationer:
More
Specifications
Cat. No.515-0093ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0913ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0963ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Polycap™ TF 75, PTFE0,2 µmMNPT/Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items1.580,00 krSpecifikationer:
More
Specifications
Cat. No.516-4453ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Polycap™ TF 75, PTFE0,2 µm1000 cm²Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items1.450,00 krSpecifikationer:
More
Specifications
Cat. No.515-0094ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Polycap™ TF 75, PTFE, sterile0,2 µmStepped barb for 10 - 12 mm tubingLog ind for tilgængelighed1 * 1 Items1.380,00 krSpecifikationer:
More
Specifications
Cat. No.516-0917ModelPolycap™ TF 75, PTFE, sterileFiltreringsareal1000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0964ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Polycap™ TF 75, PTFE0,45 µm1000 cm²Stepped barb for 6 - 10 mm tubingLog ind for tilgængelighed1 * 1 Items1.440,00 krSpecifikationer:
More
Specifications
Cat. No.515-0095ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Polycap™ TF 75, PTFE1,0 µm1000 cm²Stepped barb for 10 - 12 mm tubingLog ind for tilgængelighed1 * 1 Items1.440,00 krSpecifikationer:
More
Specifications
Cat. No.515-0096ModelPolycap™ TF 75, PTFEFiltreringsareal1000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0973ModelPolycap™ TF 150, PTFEFiltreringsareal2000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0974ModelPolycap™ TF 150, PTFEFiltreringsareal2000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0975ModelPolycap™ TF 150, PTFEFiltreringsareal2000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0976ModelPolycap™ TF 150, PTFEFiltreringsareal2000 cm²Maks. tryk4,1 bar - Specifikationer:
More
Specifications
Cat. No.516-0977ModelPolycap™ TF 150, PTFEFiltreringsareal2000 cm²Maks. tryk4,1 bar