Lead in powdered paint, certified reference material, Supelco®
Lead in powdered paint (TiO₂ matrix), Pb, 30 g
Leverandør: Merck
Lead in powdered paint, certified reference material, Supelco®
Katalog # SQC074-30G
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
ICP and AAS suitable.
Values of analyte vary lot to lot.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:30 g
- Beskrivelse:Lead in powdered paint (TiO₂ matrix)
- Symbol:Pb