Orders LinkContactUsLinkComponent
Nebulisers, PFA-ST/ST3 MicroFlow™
Nebulisers, PFA-ST/ST3 MicroFlow™
Leverandør:   PerkinElmer
Nebulisers, PFA-ST/ST3 MicroFlow™
Leverandør:  PerkinElmer
Certifikater
Nebulisers, PFA-ST/ST3 MicroFlow™
3 tilgængelige muligheder nedenfor
Product Details & Documents
The PFA-ST MicroFlow nebuliser is made from high purity, HF resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the diameter of the external capillary or probe.
  • Chemically resistant ideal for strong acids, alkalis and organics
  • Direct analysis of volatile and non-volatile organic solvents
  • Longer lifetime than glass or quartz nebulisers
  • With external 1/4 - 28 threaded connector
Exchangeable uptake capillaries allow one nebuliser to be used at different self-aspiration rates from 20 to 700 μl/min. Can be pumped from <0,1 to 3,0 ml/min.
PFA-ST3 designed for superior clog resistance and higher sensitivity compared to the standard PFA nebuliser.
PFA MicroFlow ideal for strong acids, high solids, alkalis, organic solvents, soils and high salt solutions.
Accessories information: Internal standard addition tee and FAST valve nebulizer lines are available.
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • PERCN8122192
    PFA-ST self-aspirating nebuliser
    PFA-ST self-aspirating nebuliser
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 17.000,00 kr
    17.000,00 kr
    1 * 1 Items
       17.000,00 kr
  • PERCN8145368
    PFA-ST self-aspirating nebuliser for prepFAST
    PFA-ST self-aspirating nebuliser for prepFAST
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 18.120,00 kr
    18.120,00 kr
    1 * 1 Items
       18.120,00 kr
  • PERCN8145101
    PFA-ST3 MicroFlow nebuliser
    PFA-ST3 MicroFlow nebuliser
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 20.470,00 kr
    20.470,00 kr
    1 * 1 Items
       20.470,00 kr
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • FAST valve line for PFA-ST nebuliser (0,25 mm I.Ø), green, attaches to port #3
    FAST valve line for PFA-ST nebuliser (0,25 mm I.Ø), green, attaches to port #3
    PERCN0777287
    Log ind for tilgængelighed
    1 * 1 Items - 2.190,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Internal standard addition tee for ESI ST nebuliser (0,25 mm I.Ø), green, attaches to port #3
    Internal standard addition tee for ESI ST nebuliser (0,25 mm I.Ø), green, attaches to port #3
    PERCN0777294
    Log ind for tilgængelighed
    1 * 1 Items - 9.310,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Internal standard addition tee for ESI ST nebuliser (0,5 mm I.Ø), orange, attaches to port #3
    Internal standard addition tee for ESI ST nebuliser (0,5 mm I.Ø), orange, attaches to port #3
    PERCN0777295
    Log ind for tilgængelighed
    1 * 1 Items - 9.200,00 kr