Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Detergents
VISOCOLOR® Colorimetric Test, Detergents
Leverandør:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Detergents
Leverandør:  MACHEREY-NAGEL
Certifikater
VISOCOLOR® Colorimetric Test, Detergents
2 tilgængelige muligheder nedenfor
Product Details & Documents
Reagents for the determination of detergents in water samples.
  • Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • MANA931250
  • MANA931051
  • Model:
  • MANA931250
  • MANA931051
  • Platform:
  • Colorimetric test kits
  • Colorimetric test kits
  • Opbevaringstemperatur:
  • 15...25 °C
  • 15...25 °C
  • Holdbarhed (måneder):
  • 24
  • 24
  • Vægt:
  • 415 g (Including packaging)
  • 510 g (Including packaging)
Specifikationer
Product Family Options
Produktinformation
  • Indhold
    Område
    Kemisk metode
    Til enhed
    Dimensioner
    Test pr. sæt
    Pakn
    Tilgængelighed
    Price
  • MANA931250
    VISOCOLOR® ECO Detergents anionic, refill pack
    VISOCOLOR® ECO Detergents anionic, refill pack
    Reagents for 50 tests in a box
    0,1 • 0,25 • 0,5 • 1,0 • 2,0 • 5,0 mg/L MBAS
    Methylene blue
    Visual
    235×130×45 mm (Including packaging)
    50
    50 Tests
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 627,00 kr
    627,00 kr
    1 * 1 Each
       627,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA931250
    Model
    MANA931250
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15...25 °C
    Holdbarhed (måneder)
    24
    Vægt
    415 g (Including packaging)
  • MANA931051
    VISOCOLOR® ECO Cationic detergents
    VISOCOLOR® ECO Cationic detergents
    Reagents for 50 tests, color chart and accessories in a box
    0 • 1 • 3 • 5 • 10 • 15 • 20 mg/L CTAB
    Bromophenol blue
    Visual
    239×135×47 mm (Including packaging)
    50
    1
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.300,00 kr
    1.300,00 kr
    1 * 1 Items
       1.300,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA931051
    Model
    MANA931051
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15...25 °C
    Holdbarhed (måneder)
    24
    Vægt
    510 g (Including packaging)