VISOCOLOR® Colorimetric Test, Phosphate
Supplier: MACHEREY-NAGEL
About this item
Reagents for the determination of phosphate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
7 Options Available Below
Product Details & Documents
Reagents for the determination of phosphate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- MANA920080
- MANA920082
- MANA931084
- MANA935079
- MANA920180
- MANA920182
- MANA931284
- Platform:
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Opbevaringstemperatur:
- 15...25 °C
- 15...25 °C
- 15...25 °C
- 15...25 °C
- 15...25 °C
- 15...25 °C
- 15...25 °C
- Holdbarhed (måneder):
- 24
- 24
- 36
- 24
- 24
- 24
- 36
- Vægt:
- 357,1 g (Including packaging)
- 485,22 g (Including packaging)
- 212,85 g (Including packaging)
- 99,6 g (Including packaging)
- 227 g (Including packaging)
- 363,54 g (Including packaging)
- 129 g (Including packaging)
Specifications
Product Family Options
Test kits
Product Information
- BeskrivelseIndholdOmrådeKemisk metodeTil enhedDimensionerTest pr. sætPaknAvailabilityPrice
- VISOCOLOR® HE Phosphate (DEV)Reagents for 100 tests, color chart and accessories in a box0,0 • 0,01 • 0,02 • 0,03 • 0,05 • 0,07 • 0,10 • 0,15 • 0,20 • 0,25 mg/L (PO₄ - P)Phosphomolybdenum blueVisual239×135×47 mm (Including packaging)1001 KITLog in for availability1 * 1 Each1.190,00 krSpecifications:
More
Specifications
Cat. No.MANA920080PlatformColorimetric test kitsOpbevaringstemperatur15...25 °CHoldbarhed (måneder)24Vægt357,1 g (Including packaging) - VISOCOLOR® HE PhosphateReagents for 300 tests, color chart and accessories in a box0,0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,3 • 0,4 • 0,6 • 0,8 • 1,0 mg/L (PO₄ - P)Phosphomolybdenum blueVisual239×135×47 mm (Including packaging)3001 KITLog in for availability1 * 1 Each1.210,00 krSpecifications:
More
Specifications
Cat. No.MANA920082PlatformColorimetric test kitsOpbevaringstemperatur15...25 °CHoldbarhed (måneder)24Vægt485,22 g (Including packaging) - VISOCOLOR® ECO Phosphate, test no. 5-84; Measuring range on PF-12Plus. Range on other photometers can be different.Reagents for 80 tests, color chart and accessories in a box0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 • 5 mg/L (PO₄ - P); 0,2 - 5,0 mg/L (PO₄ - P)Phosphomolybdenum bluePF-12Plus;PF-3 Soil;Advance;PF-3 Fish;Visual170×125×48 mm (Including packaging)801 KITLog in for availability1 * 1 Each547,00 krSpecifications:
More
Specifications
Cat. No.MANA931084PlatformColorimetric test kitsOpbevaringstemperatur15...25 °CHoldbarhed (måneder)36Vægt212,85 g (Including packaging) - VISOCOLOR® alpha PhosphateReagents for 70 tests and color chart in a blister2 • 5 • 10 • 15 • 20 mg/L (PO₄³⁻)Phosphomolybdenum blueVisual154×93×35 mm (Including packaging)701 KITLog in for availability1 * 1 Each412,00 krSpecifications:
More
Specifications
Cat. No.MANA935079PlatformColorimetric test kitsOpbevaringstemperatur15...25 °CHoldbarhed (måneder)24Vægt99,6 g (Including packaging)
Refill packs
Product Information
- BeskrivelseIndholdOmrådeKemisk metodeTil enhedDimensionerTest pr. sætPaknAvailabilityPrice
- VISOCOLOR® HE Phosphate (DEV), refill packReagents for 100 tests in a box0,0 • 0.01 • 0.02 • 0.03 • 0.05 • 0.07 • 0.10 • 0.15 • 0.20 • 0.25 mg/L (PO₄ - P)Phosphomolybdenum blueVisual230×130×45 mm (Including packaging)1001 KITLog in for availability1 * 1 Each736,00 krSpecifications:
More
Specifications
Cat. No.MANA920180PlatformColorimetric test kitsOpbevaringstemperatur15...25 °CHoldbarhed (måneder)24Vægt227 g (Including packaging) - VISOCOLOR® HE Phosphate, refill packReagents for 300 tests in a box0,0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,3 • 0,4 • 0,6 • 0,8 • 1,0 mg/L (PO₄ - P)Phosphomolybdenum blueVisual235×130×47 mm (Including packaging)3001 KITLog in for availability1 * 1 Each745,00 krSpecifications:
More
Specifications
Cat. No.MANA920182PlatformColorimetric test kitsOpbevaringstemperatur15...25 °CHoldbarhed (måneder)24Vægt363,54 g (Including packaging) - VISOCOLOR® ECO Phosphate, refill pack, test no. 5-84; Measuring range on PF-12Plus. Range on other photometers can be different.Reagents for 80 tests in a box0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 • 5 mg/L (PO₄ - P); 0,2 - 5,0 mg/L (PO₄ - P)Phosphomolybdenum bluePF-12Plus;PF-3 Soil;Advance;PF-3 Fish;Visual170×115×70 mm (Including packaging)801 KITLog in for availability1 * 1 Each317,00 krSpecifications:
More
Specifications
Cat. No.MANA931284PlatformColorimetric test kitsOpbevaringstemperatur15...25 °CHoldbarhed (måneder)36Vægt129 g (Including packaging)