Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, pH
VISOCOLOR® Colorimetric Test, pH
Leverandør:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, pH
Leverandør:  MACHEREY-NAGEL
Certifikater
VISOCOLOR® Colorimetric Test, pH
6 tilgængelige muligheder nedenfor
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • MANA931066
  • MANA920074
  • MANA935075
  • MANA931266
  • MANA920174
  • MANA931270
  • Model:
  • Hazardous material: Yes
  • Hazardous material: Yes
  • Hazardous material: Yes
  • Hazardous material: Yes
  • Hazardous material: Yes
  • Hazardous material: No
  • Platform:
  • Colorimetric test kits
  • Colorimetric test kits
  • Colorimetric test kits
  • Colorimetric test kits
  • Colorimetric test kits
  • Photometric test kits
  • Opbevaringstemperatur:
  • 15…25 °C
  • 15…25 °C
  • 15…25 °C
  • 15…25 °C
  • 15…25 °C
  • 15…25 °C
  • Holdbarhed (måneder):
  • 36
  • 24
  • 36
  • 36
  • 24
  • 18
  • Vægt:
  • 205 g (including packaging)
  • 277,22 g (including packaging)
  • 53,3 g (including packaging)
  • 116 g (including packaging)
  • 142 g (including packaging)
  • 105,91 g (including packaging)
Specifikationer
Product Family Options
Test Kit
Produktinformation
  • Indhold
    Område
    Kemisk metode
    Til enhed
    Dimensioner
    Model
    Test pr. sæt
    Pakn
    Tilgængelighed
    Price
  • MANA931066
    VISOCOLOR® ECO pH 4 - 9
    VISOCOLOR® ECO pH 4 - 9
    Reagents for 450 tests, color chart and accessories in a box
    4,0 • 5,0 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0
    Mixed indicator
    Visual
    168×112×40 mm (including packaging)
    Hazardous material: Yes
    450
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 557,00 kr
    557,00 kr
    1 * 1 Each
       557,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA931066
    Model
    Hazardous material: Yes
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15…25 °C
    Holdbarhed (måneder)
    36
    Vægt
    205 g (including packaging)
  • MANA920074
    VISOCOLOR® HE pH 4 - 10
    VISOCOLOR® HE pH 4 - 10
    Reagents for 500 tests, color chart and accessories in a box
    4,0 • 5,0 • 5,5 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0 • 10,0
    Mixed indicator
    Visual
    239×135×47 mm (including packaging)
    Hazardous material: Yes
    500
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 1.190,00 kr
    1.190,00 kr
    1 * 1 Each
       1.190,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA920074
    Model
    Hazardous material: Yes
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15…25 °C
    Holdbarhed (måneder)
    24
    Vægt
    277,22 g (including packaging)
  • MANA935075
    VISOCOLOR® alpha pH 5 - 9
    VISOCOLOR® alpha pH 5 - 9
    Reagents for 200 tests and color chart in a blister
    5.0 • 5.5 • 6.0 • 6.5 • 7.0 • 7.5 • 8.0 • 8.5 • 9.0 pH
    Mixed indicator
    Visual
    154×93×35 mm (including packaging)
    Hazardous material: Yes
    200
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 338,00 kr
    338,00 kr
    1 * 1 Each
       338,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA935075
    Model
    Hazardous material: Yes
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15…25 °C
    Holdbarhed (måneder)
    36
    Vægt
    53,3 g (including packaging)
Refill packs
Produktinformation
  • Indhold
    Område
    Kemisk metode
    Til enhed
    Dimensioner
    Model
    Test pr. sæt
    Pakn
    Tilgængelighed
    Price
  • MANA931266
    VISOCOLOR® ECO pH 4 - 9, refill pack
    VISOCOLOR® ECO pH 4 - 9, refill pack
    Reagents for 450 tests in a box
    4,0 • 5,0 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0
    Mixed indicator
    Visual
    170×115×70 mm (including packaging)
    Hazardous material: Yes
    450
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 264,00 kr
    264,00 kr
    1 * 1 Each
       264,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA931266
    Model
    Hazardous material: Yes
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15…25 °C
    Holdbarhed (måneder)
    36
    Vægt
    116 g (including packaging)
  • MANA920174
    VISOCOLOR® HE pH 4 - 10, refill pack
    VISOCOLOR® HE pH 4 - 10, refill pack
    Reagents for 500 tests in a box
    4,0 • 5,0 • 5,5 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0 • 10,0
    Mixed indicator
    Visual
    237×130×45 mm (including packaging)
    Hazardous material: Yes
    500
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 736,00 kr
    736,00 kr
    1 * 1 Each
       736,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA920174
    Model
    Hazardous material: Yes
    Platform
    Colorimetric test kits
    Opbevaringstemperatur
    15…25 °C
    Holdbarhed (måneder)
    24
    Vægt
    142 g (including packaging)
  • MANA931270
    VISOCOLOR® ECO pH 6,1 - 8,4, refill pack, test no.5-70; Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation
    VISOCOLOR® ECO pH 6,1 - 8,4, refill pack, test no.5-70; Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation
    Reagents for 150 tests in a box
    pH: 6.1 - 8.4
    Mixed indicator
    PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance
    170×115×70 mm (including packaging)
    Hazardous material: No
    150
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 688,00 kr
    688,00 kr
    1 * 1 Each
       688,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA931270
    Model
    Hazardous material: No
    Platform
    Photometric test kits
    Opbevaringstemperatur
    15…25 °C
    Holdbarhed (måneder)
    18
    Vægt
    105,91 g (including packaging)