Orders LinkContactUsLinkComponent
LED excitation modules
LED excitation modules
Leverandør:   MOTIC
LED excitation modules
Leverandør:  MOTIC
Certifikater
LED excitation modules
LED excitation modules
3 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • MOTI1101011400251
    LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
    LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 13.700,00 kr
    13.700,00 kr
    1 * 1 Items
       13.700,00 kr
  • MOTI1101011400241
    LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
    LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 11.930,00 kr
    11.930,00 kr
    1 * 1 Items
       11.930,00 kr
  • MOTI1101011400231
    LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 11.930,00 kr
    11.930,00 kr
    1 * 1 Items
       11.930,00 kr