Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Molybdenum
QUANTOFIX® Test Strips, Molybdenum
QUANTOFIX® lubricheck, 0 • 15 • 50 • 75 • 130 • 200 - mmol/L - KOH, Color reaction with pH indicator, 40×40×120 mm (packaging), 100 Tests, 100 Tests
Katalog # MANA91336
Leverandør:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Molybdenum
Katalog # MANA91336
Leverandør:  MACHEREY-NAGEL
CAS nummer:  
QUANTOFIX® Test Strips, Molybdenum
QUANTOFIX® Test Strips, Molybdenum
QUANTOFIX® lubricheck, 0 • 15 • 50 • 75 • 130 • 200 - mmol/L - KOH, Color reaction with pH indicator, 40×40×120 mm (packaging), 100 Tests, 100 Tests
CAS nummer:  
Price
387,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of molybdenum in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 1 minute one gets a reliable result.
Specifikationer
Specifikationer
  • Kemisk metode:
    Color reaction with pH indicator
  • Pakn:
    100 Tests
  • Beskrivelse:
    QUANTOFIX® lubricheck
  • Dimensioner:
    40×40×120 mm (packaging)
  • Test pr. sæt:
    100 Tests
  • Område:
    0 • 15 • 50 • 75 • 130 • 200 - mmol/L - KOH
  • Indhold:
    100 test strips in a tube, instruction leaflet
  • Cat. No.:
    MANA91336
  • Overensstemmelse:
    No hazardous material
  • Platform:
    Semi-quantitative test strips
  • Holdbarhed (måneder):
    30 (from production)
  • Vægt:
    44 g
  • Instrument:
    No
  • Parameter:
    All reagents included
  • Method:
    Cooling lubricants - Mixed indicator (yellow → blue)