QUANTOFIX® Test Strips, Arsenic
Om denne vare
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale
- Always ready to use: No accessories necessary for evaluation
- Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
The test strips allow the quick and easy determination of arsenic in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 20 to 30 minutes one gets reliable results.
As a naturally occurring element, arsenic is widely distributed in the earth's crust. In the natural environment it is most frequently present as inorganic arsenic compound in combination with sulphur or oxygen. Organic arsenic compounds can be used as pesticides. Arsenic is toxic and causes skin diseases, keratosis and melanomas. Therefore, arsenic levels in drinking water have to be monitored thoroughly.
Delivery information: QUANTOFIX® Arsenic Sensitive kit is supplied with a rugged case, tube with 100 test strips, reagents and all necessary equipment
3 tilgængelige muligheder nedenfor
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale
- Always ready to use: No accessories necessary for evaluation
- Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
The test strips allow the quick and easy determination of arsenic in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 20 to 30 minutes one gets reliable results.
As a naturally occurring element, arsenic is widely distributed in the earth's crust. In the natural environment it is most frequently present as inorganic arsenic compound in combination with sulphur or oxygen. Organic arsenic compounds can be used as pesticides. Arsenic is toxic and causes skin diseases, keratosis and melanomas. Therefore, arsenic levels in drinking water have to be monitored thoroughly.
Delivery information: QUANTOFIX® Arsenic Sensitive kit is supplied with a rugged case, tube with 100 test strips, reagents and all necessary equipment
Documents
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- MANA91334
- MANA91332
- MANA91345
- Method:
- Arsenic - Modified Gutzeit test (white → yellow-brown)
- Arsenic - Modified Gutzeit test (white → yellow-brown)
- Arsenic - Modified Gutzeit test (white → yellow-brown)
- Parameter:
- Requires additional reagent
- Requires additional reagent
- Requires additional reagent
- Overensstemmelse:
- Hazardous material
- Hazardous material
- Hazardous material
- Indhold:
- 100 test strips in a tube, additional regaents, instruction leaflet in a box
- 100 test strips in a tube, additional regaents, instruction leaflet in a box
- 100 test strips in a tube, additional regaents, instruction leaflet in a box
- Instrument:
- No
- No
- No
- Platform:
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Holdbarhed (måneder):
- 30 (from production)
- 30 (from production)
- 30 (from production)
- Vægt:
- 435,31 g
- 293,04 g
- 1162,9 g
Specifikationer
Product Family Options
Produktinformation
- OmrådeKemisk metodeDimensionerTest pr. sætPaknTilgængelighedPrice
- MANA91334QUANTOFIX Arsenic 10QUANTOFIX Arsenic 100 • 0.01 • 0.025 • 0.05 • 0.1 • 0.5 - mg/L - As³⁺ ⁵⁺Colour reaction with mercury bromide185×145×47 mm (package)100 test(s)100 TestsLog ind for tilgængelighed1 * 1 Each900,00 krSpecifikationer:
More
Specifications
Cat. No.MANA91334MethodArsenic - Modified Gutzeit test (white → yellow-brown)ParameterRequires additional reagentOverensstemmelseHazardous materialIndhold100 test strips in a tube, additional regaents, instruction leaflet in a boxInstrumentNoPlatformSemi-quantitative test stripsHoldbarhed (måneder)30 (from production)Vægt435,31 g - MANA91332QUANTOFIX Arsenic 50QUANTOFIX Arsenic 500 • 0.05 • 0.1 • 0.5 • 1.0 • 1.7 • 3.0 - mg/L - As³⁺ ⁵⁺Colour reaction with mercury bromide168×112×40 mm (package)100 test(s)100Log ind for tilgængelighed1 * 100 Items857,00 krSpecifikationer:
More
Specifications
Cat. No.MANA91332MethodArsenic - Modified Gutzeit test (white → yellow-brown)ParameterRequires additional reagentOverensstemmelseHazardous materialIndhold100 test strips in a tube, additional regaents, instruction leaflet in a boxInstrumentNoPlatformSemi-quantitative test stripsHoldbarhed (måneder)30 (from production)Vægt293,04 g - MANA91345QUANTOFIX Arsenic SensitiveQUANTOFIX Arsenic Sensitive0 • 0.005 • 0.01 • 0.025 • 0.05 • 0.1 • 0.25 • 0.5 - mg/L - As³⁺ ⁵⁺Colour reaction with mercury bromide340×280×85 mm (package)100 test(s)1 SETLog ind for tilgængelighed1 * 1 Each1.980,00 krSpecifikationer:
More
Specifications
Cat. No.MANA91345MethodArsenic - Modified Gutzeit test (white → yellow-brown)ParameterRequires additional reagentOverensstemmelseHazardous materialIndhold100 test strips in a tube, additional regaents, instruction leaflet in a boxInstrumentNoPlatformSemi-quantitative test stripsHoldbarhed (måneder)30 (from production)Vægt1162,9 g