VISOCOLOR® ECO Fish analysis case
Leverandør: MACHEREY-NAGEL
Om denne vare
The VISOCOLOR Fish analysis kit for water quality testing.
- Specially developed to meet the requirements for testing fishing waters
It combines reagents and accessories for the 6 most important water parameters: ammonium, nitrate, nitrite, phosphate, pH and total hardness.
4 tilgængelige muligheder nedenfor
Product Details & Documents
The VISOCOLOR Fish analysis kit for water quality testing.
- Specially developed to meet the requirements for testing fishing waters
It combines reagents and accessories for the 6 most important water parameters: ammonium, nitrate, nitrite, phosphate, pH and total hardness.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- MANA933101
- MANA933151
- MANA933201
- MANA933301
- Model:
- Hazardous material: Yes
- Hazardous material: No
- Hazardous material: Yes
- Hazardous material: No
- Platform:
- —
- Accessories
- —
- Accessories
- Opbevaringstemperatur:
- 15...25 °C
- —
- 15...25 °C
- —
- Holdbarhed (måneder):
- 18
- —
- 18
- —
- Vægt:
- 1381,5 g (Including packaging)
- 69,9 g (Including packaging)
- 455 g (Including packaging)
- 36,3 g (Including packaging)
Specifikationer
Product Family Options
Produktinformation
- IndholdDimensionerModelPaknTilgængelighedPrice
- MANA933101VISOCOLOR® FISH reagent caseVISOCOLOR® FISH reagent caseCase, test kits, accessories, instructions325×238×155 mm (Including packaging)Hazardous material: Yes1 SETLog ind for tilgængelighed1 * 1 Each1.200,00 krSpecifikationer:
More
Specifications
Cat. No.MANA933101ModelHazardous material: YesPlatform—Opbevaringstemperatur15...25 °CHoldbarhed (måneder)18Vægt1381,5 g (Including packaging) - MANA933151VISOCOLOR® FISH reagent case manualVISOCOLOR® FISH reagent case manual1 manual270×180×3 mm (Including packaging)Hazardous material: No1Log ind for tilgængelighed1 * 1 Items208,00 krSpecifikationer:
More
Specifications
Cat. No.MANA933151ModelHazardous material: NoPlatformAccessoriesOpbevaringstemperatur—Holdbarhed (måneder)—Vægt69,9 g (Including packaging) - MANA933201VISOCOLOR® FISH reagent caseVISOCOLOR® FISH reagent caseReagents, spoon195×170×50 mm (Including packaging)Hazardous material: Yes1 SETLog ind for tilgængelighed1 * 1 Each685,00 krSpecifikationer:
More
Specifications
Cat. No.MANA933201ModelHazardous material: YesPlatform—Opbevaringstemperatur15...25 °CHoldbarhed (måneder)18Vægt455 g (Including packaging) - MANA933301VISOCOLOR® FISH reagent case-color scaleVISOCOLOR® FISH reagent case-color scale1 colour chart330×200×2 mm (Including packaging)Hazardous material: No1Log ind for tilgængelighed1 * 1 Items162,00 krSpecifikationer:
More
Specifications
Cat. No.MANA933301ModelHazardous material: NoPlatformAccessoriesOpbevaringstemperatur—Holdbarhed (måneder)—Vægt36,3 g (Including packaging)