HPLC columns, SUPELCOSIL™, LC-8-DB, Supelco®
Supplier: Merck
Certificates
About this item
These SUPELCOSIL LC-DB phases are specially deactivated for basic compounds. These columns provide less retention, better peak shape, and higher efficiency for organic bases than can be obtained on conventional reversed-phase columns.
Delivery: Supelguard cartridge kit includes one cartridge, a stand-alone holder, a piece of tubing, and two nuts and ferrules.
Specifications
- Catalog No:
- 59563.
- 58354.
- 58991.
- 58347.
- 58991C40
- 58976.
- 58344.
- 59553.
- 58354C40
- 58990-U
- Kulstofbelastning:
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- Endcapped:
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Surface area:
- 170 m²/g
- 170 m²/g
- 170 m²/g
- 170 m²/g
- 170 m²/g
- 170 m²/g
- 170 m²/g
- 170 m²/g
- 170 m²/g
- 170 m²/g
- USP code:
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7