Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Flubi-2 fluorescent pH indicator

Flubi-2 fluorescent pH indicator

5 mg
Katalog # 51024.
Leverandør:   Biotium
Flubi-2 fluorescent pH indicator
Katalog # 51024.
Leverandør:  Biotium
CAS nummer:  
Flubi-2 fluorescent pH indicator
Flubi-2 fluorescent pH indicator

5 mg

CAS nummer:  
Price
2.560,00 kr
Log ind for tilgængelighed
1 * 5 mg
Product Details & Documents

Flubi-2 is the hydrolyzed product of the site-directed fluorogenic probe Flubida-2 (#51022) and can be used as a standard for calibration. The dye can also be microinjected into cells to probe the local pH where avidin fusion proteins are located.

  • Cell permeability: Membrane impermeant
  • pKa 6.5
  • Colors: Green
  • Yellow solid soluble in DMSO
  • Store at -20°C
  • Formel:C₃₉H₄₄N₄O₈S
  • Molecular Weight:728.85 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    5 mg

Specifikationer