Supelco® TraceCERT® Organic Reference Standard, Toluene solution
Toluene solution, 200 μg/ml in methanol, Certified reference material, TraceCERT®, Ampule of 1 ml, 1 Ampoul
Katalog # CRM48620
Leverandør: Merck
Supelco® TraceCERT® Organic Reference Standard, Toluene solution
Katalog # CRM48620
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
Advarsel: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311
Specifikationer
Specifikationer
- Koncentration:200 μg/ml in methanol
- Pakn:1 Ampoul
- Standard:Certified reference material, TraceCERT®
- Beskrivelse:Toluene solution
- Pk Info:Ampule of 1 ml
- Cat. No.:CRM48620
- M-Clarity quality level:MQ100
- CAS-nr:108-88-3
- Fareklassebeskrivelse:H225 - H301 + H311 + H331 - H370
- Flammepunkt:11 °C
- Opbevaring betingelser:2...30 °C
- Synonym(s):Toluene solution
- Shipping conditions:STD