Supelco® TraceCERT® Organic Reference Standard, Benzo[a]pyrene solution
Benzo[a]pyrene solution, 1000 μg/ml in acetone, Certified reference material, TraceCERT®, 1 mL
Katalog # CRM40071
Leverandør: Merck
Supelco® TraceCERT® Organic Reference Standard, Benzo[a]pyrene solution
Katalog # CRM40071
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS08, GHS07
Advarsel: P202 - P210 - P233 - P273 - P305 + P351 + P338 - P308 + P313
Specifikationer
Specifikationer
- Koncentration:1000 μg/ml in acetone
- Pakn:1 mL
- Standard:Certified reference material, TraceCERT®
- Beskrivelse:Benzo[a]pyrene solution
- Cat. No.:CRM40071
- M-Clarity quality level:MQ100
- CAS-nr:50-32-8
- Molekylvægt:252,31
- MDL No.:MFCD00003602
- Fareklassebeskrivelse:H225 - H319 - H336 - H340 - H350 - H412
- Flammepunkt:–17,0 °C – Closed cup
- Opbevaring betingelser:2…8 °C
- Shipping conditions:STD



