Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, beta-Myrcene solution

Supelco® Organic Reference Standard, beta-Myrcene solution

beta-Myrcene solution, 2000 µg/ml in hexane, Certified reference material, Ampule of 1 ml, 1 mL
Katalog # CRM42262
Leverandør:   Merck
Supelco® Organic Reference Standard, beta-Myrcene solution
Katalog # CRM42262
Leverandør:  Merck
CAS nummer:  
Supelco® Organic Reference Standard, beta-Myrcene solution

beta-Myrcene solution, 2000 µg/ml in hexane, Certified reference material, Ampule of 1 ml, 1 mL

CAS nummer:  
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 1 mL
Product Details & Documents
  • GHS signal: GHS02, GHS08, GHS07, GHS09

Caution: P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331

Specifikationer

Specifikationer

  • Koncentration:
    2000 µg/ml in hexane
  • Pakn:
    1 mL
  • Standard:
    Certified reference material
  • Beskrivelse:
    beta-Myrcene solution
  • Pk Info:
    Ampule of 1 ml
  • Cat. No.:
    CRM42262
  • M-Clarity quality level:
    MQ300
  • CAS-nr:
    123-35-3
  • Fareklassebeskrivelse:
    H225 - H304 - H315 - H336 - H361f - H373 - H411
  • Flammepunkt:
    –26,0 °C – Closed cup
  • Opbevaring betingelser:
    2…8 °C
  • Shipping conditions:
    WICE

Specifikationer