Supelco® Organic Reference Standard, (+)-Menthol solution
(+)-Menthol solution, 2000 µg/ml in methanol, Certified reference material, Ampule of 1 ml, 1 mL
Katalog # CRM40474
Leverandør: Merck
Supelco® Organic Reference Standard, (+)-Menthol solution
Katalog # CRM40474
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311
Specifikationer
Specifikationer
- Koncentration:2000 µg/ml in methanol
- Pakn:1 mL
- Standard:Certified reference material
- Beskrivelse:(+)-Menthol solution
- Pk Info:Ampule of 1 ml
- Cat. No.:CRM40474
- M-Clarity quality level:MQ300
- CAS-nr:15356-60-2
- Fareklassebeskrivelse:H225 - H301 + H311 + H331 - H370
- Flammepunkt:9,7 °C - Closed cup
- Opbevaring betingelser:2…8 °C
- Shipping conditions:WICE