Supelco® Organic Reference Standard, (-)-Guaiol solution
(-)-Guaiol solution, 2000 µg/ml in methanol, Certified reference material, 1 mL
CRM40917
: Merck
- GHS signal: GHS02, GHS06, GHS08
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311
- Koncentration:2000 µg/ml in methanol
- Pakn:1 mL
- Standard:Certified reference material
- Beskrivelse:(-)-Guaiol solution
- Cat. No.:CRM40917
- M-Clarity quality level:MQ300
- Fareklassebeskrivelse:H225 - H301 + H311 + H331 - H370
- Flammepunkt:9,7 °C - Closed cup
- Opbevaring betingelser:2…8 °C
- Synonym(s):Cannabis terpenes; Terpenes
- Shipping conditions:WICE
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



