Drug and metabolite standards, (+/-)-propranolol-D7 (ring-D7) solution, (CRM), Supelco®
Katalog # P-085-1ML
Leverandør: Merck
CAS nummer:
Specifikationer
- Koncentration:100 µg/ml in methanol with 5% 1 M HCl
- Pakn:1 mL
- Standard:Certified reference material
- CAS-nr:344298-99-3
- Beskrivelse:(+/-)-Propranolol-D7 (ring-D7) solution
- Environmentally Preferable:
- Beskrivelse:(+/-)-Propranolol-D7 (ring-D7) solution
- Pk Info:ampoule of 1 ml