Orders LinkContactUsLinkComponent
Peroxidase substrates, TrueBlue™
Peroxidase substrates, TrueBlue™
Peroxidase substrates, TrueBlue™
Leverandør:  SERACARE LIFE SCIENCES
Certifikater
Peroxidase substrates, TrueBlue™
2 tilgængelige muligheder nedenfor
Product Details & Documents
KPL TrueBlue™ peroxidase substrate is a highly sensitive chromagenic substrate for visualisation of horseradish peroxidase-labeled reporter reagents.
  • Provides a brilliant blue specific stain which provides excellent contrast with red counterstains
  • Free of known carcinogens
  • Single component, ready to use, liquid substrate
  • No mixing is required
KPL TrueBlue™ peroxidase substrate is a buffered solution containing 3,3',5,5'-tetramethylbenzidine (TMB) and H2O2. It is optimised for use with cellular preparations and can be used for immunoblotting procedures, but not for microwell ELISA or other applications requiring a soluble reaction product.
Caution: Store at room temperature (22 to 28 °C). TrueBlue is stable for up to 30 months from the date of manufacture.
Documents
Product Family Options
Produktinformation
  • Pakn
    Tilgængelighed
    Price
  • KPLI5510-0052
    TrueBlue™ peroxidase substrate, 1×10 ml, sufficient to stain approximately 25 - 50 slides
    TrueBlue™ peroxidase substrate, 1×10 ml, sufficient to stain approximately 25 - 50 slides
    10 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 mL
    1 * 10 mL
  • KPLI5510-0030
    TrueBlue™ peroxidase substrate, 2×100 ml, sufficient to stain approximately 500 - 1000 slides
    TrueBlue™ peroxidase substrate, 2×100 ml, sufficient to stain approximately 500 - 1000 slides
    2
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 4.890,00 kr
    4.890,00 kr
    1 * 2 Items
       4.890,00 kr