Heat resistant gloves, uvex k-basic extra
Supplier: UVEX
About this item
These coarse-knitted gloves made of 100% Kevlar® are lined with cotton, making them ideal for protecting the wearer against heat and cuts.
- Temperature resistant up to +250 °C
- Excellent protection against cuts
- Breathable
- Very good thermal insulation in direct contact with warm to hot objects
- Good wearer comfort with the cotton lining on the inside
Ideal for welding applications, metal processing, automotive industry, glass industry and foundries.
Certifications: EN 388:2016 (2442D), EN 407 (X2XXXX)
3 Options Available Below
Product Details & Documents
These coarse-knitted gloves made of 100% Kevlar® are lined with cotton, making them ideal for protecting the wearer against heat and cuts.
- Temperature resistant up to +250 °C
- Excellent protection against cuts
- Breathable
- Very good thermal insulation in direct contact with warm to hot objects
- Good wearer comfort with the cotton lining on the inside
Ideal for welding applications, metal processing, automotive industry, glass industry and foundries.
Certifications: EN 388:2016 (2442D), EN 407 (X2XXXX)
Documents
UVEX6017904_TDS_EN
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- UVEX6017902
- UVEX6017904
- UVEX6017905
- Model:
- 6658
- 6658
- 6658
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Foringsmateriale:
- Bomuld
- Bomuld
- Bomuld
- Måler:
- 7
- 7
- 7
- Manchet:
- Strikket
- Strikket
- Strikket
- EN norm:
- EN 388:2016 (2442D), EN 407 (X2XXXX)
- EN 388:2016 (2442D), EN 407 (X2XXXX)
- EN 388:2016 (2442D), EN 407 (X2XXXX)
Specifications
Product Family Options
Product Information
- Handske størrelseFarveLængdeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.UVEX6017902Model6658Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeBomuldMåler7ManchetStrikketEN normEN 388:2016 (2442D), EN 407 (X2XXXX) - Specifications:
More
Specifications
Cat. No.UVEX6017904Model6658Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeBomuldMåler7ManchetStrikketEN normEN 388:2016 (2442D), EN 407 (X2XXXX) - Specifications:
More
Specifications
Cat. No.UVEX6017905Model6658Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeBomuldMåler7ManchetStrikketEN normEN 388:2016 (2442D), EN 407 (X2XXXX)