Furnace tong, Ward pattern
590 mm, Furnace tong
Katalog # RLAA5704
Leverandør: R L ENTERPRISES
Gå til:
Product Details & Documents
This tong is used for protection and safety when handling heated objects in the laboratory.
- Manufactured from stainless steel
- For handling a wide variety of crucibles, especially in muffle furnaces
- Operated by thumb action with one hand
Fitted with a circular shield to protect the hand against heat from the furnace.
Specifikationer
- Længde:590 mm
- Model:Furnace tong