Replacement rotor seals for Rheodyne™ Injector models 7000/7010/7040
Catalog # HYPE7010-071
Supplier: Thermo Fisher Scientific
Specifications
- Til:Rheodyne
- Beskrivelse:Tefzel Rotor Seal for 7010
- Pakn:1
Specifications
About this item
Thermo Scientific™ Rheodyne™ replacement rotor seals for injectors made of ETFE to maintain the performance of Rheodyne™ sample injectors for high-performance liquid chromatography (HPLC).