Orders LinkContactUsLinkComponent
Lids for Clifton Baths
Lids for Clifton Baths
Supplier:   NICKEL ELECTRO
Lids for Clifton Baths
Supplier:  NICKEL ELECTRO
Certificates
Lids for Clifton Baths
Lids for Clifton Baths
11 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Beskrivelse
    Til
    Availability
    Price
  • Clear plastic lift-off gabled lid designed to allow full use of the whole width of a Clifton water bath with the lowest height of the gable being 45 mm above the bath. This means test tube racks or flasks can be placed across the whole width of the bath without fouling the lid, maximising the bath working area. Insulated handle. Lid fits into the tank ensuring all condensation runs back into the bath and not down the side onto the bench top.
    NE2-10D, NE1-8, NE2D-4/8, NE2-8D, NE5-10D, NE2D-8/22 and NE1D-8
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.380,00 kr
    1.380,00 kr
    1 * 1 Items
       1.380,00 kr
  • Låg af rustfrit stål med gavle
    14-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.570,00 kr
    2.570,00 kr
    1 * 1 Items
       2.570,00 kr
  • Hængslet låg af rustfrit stål med gavle
    22- og 28-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.920,00 kr
    6.920,00 kr
    1 * 1 Items
       6.920,00 kr
  • Låg af rustfrit stål med gavle
    22- og 28-liters bade (NE1/NE1B/NE2D/NE5 og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.050,00 kr
    3.050,00 kr
    1 * 1 Items
       3.050,00 kr
  • Låg af rustfrit stål med gavle
    4-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.410,00 kr
    2.410,00 kr
    1 * 1 Items
       2.410,00 kr
  • Låg af rustfrit stål med gavle
    8-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.700,00 kr
    2.700,00 kr
    1 * 1 Items
       2.700,00 kr
  • Låg, rustfrit stål, 4x105 mm huller med ringsæt
    14-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.510,00 kr
    3.510,00 kr
    1 * 1 Items
       3.510,00 kr
  • Låg, rustfrit stål, 6x105 mm huller med ringsæt
    22-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 4.840,00 kr
    4.840,00 kr
    1 * 1 Items
       4.840,00 kr
  • Låg, rustfrit stål, 2x105 mm huller med ringsæt
    4-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.220,00 kr
    2.220,00 kr
    1 * 1 Items
       2.220,00 kr
  • Låg, rustfrit stål, 4x83 mm huller med ringsæt
    8-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.860,00 kr
    2.860,00 kr
    1 * 1 Items
       2.860,00 kr
  • Låg, rustfrit stål, 6x81 mm huller med ringsæt
    14-liters bade (NE1/NE1B/NE2D og Duobaths)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.850,00 kr
    3.850,00 kr
    1 * 1 Items
       3.850,00 kr