Orders LinkContactUsLinkComponent
Microplate reader, SpectraMax® L
Microplate reader, SpectraMax® L
Leverandør:   Molecular Devices
Microplate reader, SpectraMax® L
Leverandør:  Molecular Devices
Certifikater
Microplate reader, SpectraMax® L
Microplate reader, SpectraMax® L
7 tilgængelige muligheder nedenfor
Product Details & Documents
The SpectraMax® L microplate reader measures flash and glow luminescence assays in both 96- and 384-well plates.
  • Detect dim samples with unsurpassed sensitivity
  • Avoid saturation issues with 9 orders of dynamic range
  • Enable robust performance with autorinse injectors
  • Ensure compliance with IQ/OQ and FDA CFR 21 Part 11 compliance tools
  • Easy set-up and analysis with industry-leading SoftMax Pro software
SpectraMax® L is upgradeable to include a barcode reader, programmable injectors, and dual-channel configurations for a 2-fold increase in medium throughput. The high sensitivity, low-noise photomultiplier tubes (PMTs), and variable-volume injector configuration make this reader ideal for fast kinetic assays requiring continuous monitoring during injection and measurement.
Dual injector option includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Dual injector option (auto aperture) includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Two PMTs with automatic switching option enables wavelength-based measurements such as bioluminescence resonance energy transfer (BRET) assays and a 2-fold increase in throughput for traditional luminescence measurements. Wavelength-based measurements require purchase of appropriate filters.
Four injector option is suitable for SpectraMax® L readers with two PMTs and includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Certifications: For full details about the options available with this system please contact your local Avantor sales office.
Delivery information: System includes SpectraMax® L luminometer for reading 96- and 384- well microplates with current version of SoftMax Pro software, AutoRange for >9 orders of dynamic range, Photon Counting for high sensitivity, landscape/portrait plate reading, incubator, user manual, USB interface cable, 1 year MDCare PhD Technical Support Plan and 1 year warranty covering parts and labour.
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • MLDVSPECTRALCONFIG
    SpectraMax® L luminescence microplate reader
    SpectraMax® L luminescence microplate reader
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Options
Produktinformation
  • Tilgængelighed
    Price
  • MLDV0310-5628
    Single PMT with manual aperture
    Single PMT with manual aperture
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • MLDV0310-5126
    Dual injector option
    Dual injector option
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • MLDV0310-5649
    Single PMT with automatic switching between 96- and 384- well micrplates
    Single PMT with automatic switching between 96- and 384- well micrplates
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • MLDV0310-5882
    Dual injector option for SpectraMax® L reader (auto aperture)
    Dual injector option for SpectraMax® L reader (auto aperture)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • MLDV0310-5650
    Dual PMTs with automatic switching between 96- and 384-well plates
    Dual PMTs with automatic switching between 96- and 384-well plates
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • MLDV0310-5694
    Four injector option for SpectraMax® L readers with two PMTs
    Four injector option for SpectraMax® L readers with two PMTs
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Complementary Products
Produktinformation
  • Katalog #
    Beskrivelse
    Sterilitet
    Serum format
    Pakn
    Anbefalet arbejdsvolumen
    Farve
    Medieformat
    Steril
    Dyrkningsområde
    Pakket
    Tilgængelighed
    Price
  • MLDV0200-6017
    40-plate stacker magazines (pair)
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr
  • MLDV0200-6016
    20-plate stacker magazines (pair)
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr
  • MLDV0200-6187
    50-plate stacker magazines (pair)
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr
  • MLDV5005331
    Loading tool for facilitating loading and unloading plates
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr
  • S1810-500
    Foetal bovine serum, South American origin
    Frossen
    15
    Log ind for tilgængelighed
    1 * 500 mL - 2.400,00 kr
  • MLDVSTAKMAX
    StakMax® microplate handling system
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr
  • 30028-L1
    120 Tests
    Log ind for tilgængelighed
    1 * 1 Each - 1.130,00 kr
  • 30081-1
    150 Assays
    Log ind for tilgængelighed
    1 * 1 Each - 2.440,00 kr
  • X0930-100
    100 mL
    11
    Log ind for tilgængelighed
    1 * 100 mL - 164,00 kr
  • 734-2097
    96 MicroWell™-plade, fladbundede brønde, med låg
    transparent
    06
    Log ind for tilgængelighed
    1 * 50 Items - 1.630,00 kr
  • L0135-500
    Flydende
    Ham's F12 with L-glutamine
    67
    Log ind for tilgængelighed
    1 * 500 mL - 117,00 kr
  • 735-0523
    Microlite™ 1+ strip, 1×12 pr. strip
    Log ind for tilgængelighed
    1 * 320 Items - 1.710,00 kr
  • 734-2327
    96-brønds plader, flad bund, TC-behandlede
    +
    0,33 cm²
    1/pose
    klar
    17
    Log ind for tilgængelighed
    1 * 100 Items - 1.940,00 kr
  • CORN3904BC
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    20/bag
    75 - 200 µl
    sort
    21
    360 µl
    Log ind for tilgængelighed
    1 * 100 Items - 9.690,00 kr