Orders LinkContactUsLinkComponent
Watch glasses
  PERCN9308003
Watch glasses
  PERCN9308003
Watch glasses
4.720,00 kr
1 * 1.000 Items
Disposable watch glasses to promote gentle refluxing. For use with 50 ml and 100 ml DigiTUBEs.