Orders LinkContactUsLinkComponent
Hobby knife
Hobby knife
150 mm, Hobby knife, incl. 3 spare blades, 34 g
Katalog # WEDO78621
Leverandør:   WEDO WERNER DORSCH
Hobby knife
Katalog # WEDO78621
Leverandør:  WEDO WERNER DORSCH
Hobby knife
Hobby knife
150 mm, Hobby knife, incl. 3 spare blades, 34 g
Price
101,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This knife has a pointed scalpel which allows fine and precise cutting of paper, cardboard, and foil.
WEDO® aluminum scalpel for graphics, work, and hobby. The rubberized grip zone ensures comfortable handling. With protective cap for safe transport and storage. Ideal for fine cutting work in paper, cardboard, and foil. Packaged in a hinged PP box with three replacement blades.
Delivery information: Includes three spare blades.
Specifikationer
  • Beskrivelse:
    Hobby knife, incl. 3 spare blades, 34 g
  • Længde:
    150 mm