Orders LinkContactUsLinkComponent
dNTP mix, 2 mM
dNTP mix, 2 mM
dNTP mix, 2 mM, 1 KIT
Katalog # R0242
Leverandør:   Thermo Fisher Scientific
dNTP mix, 2 mM
Katalog # R0242
CAS nummer:  
dNTP mix, 2 mM
dNTP mix, 2 mM
dNTP mix, 2 mM, 1 KIT
CAS nummer:  
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
dNTP mix contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 2 mM.
  • Greater than 99% purity confirmed by HPLC
  • Free of human and E. coli DNA
  • Highly stable
Suitable for used in all molecular biology applications, including PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, RDA, MDA, DNA labelling, and DNA sequencing.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    dNTP mix, 2 mM