Orders LinkContactUsLinkComponent
Shoe covers, CPE, embossed
Supplier:   VWR Collection
Shoe covers, CPE, embossed
Supplier:  VWR Collection
Certificates
Shoe covers, CPE, embossed
2 Options Available Below
Product Details & Documents
These shoe covers are made from CPE.
  • Thickness: 40 µm
  • Latex-free elastic
 : 2000 per carton.
Product Family Options
Product Information
  • Farve
    Højde
    Længde
    Availability
    Price
  • grøn
    150 mm
    410 mm
    Log in for availability
    Log in for availability
    1 * 2.000 Items
    1 * 2.000 Items
  • rød
    150 mm
    410 mm
    Log in for availability
    Log in for availability
    1 * 2.000 Items
    1 * 2.000 Items