Orders LinkContactUsLinkComponent
His·Bind® buffer kit, Novagen®
His·Bind® buffer kit, Novagen®
His·Bind® buffer kit, 1 KIT
Katalog # 69755-3
His·Bind® buffer kit, Novagen®
Katalog # 69755-3
CAS nummer:  
His·Bind® buffer kit, Novagen®
His·Bind® buffer kit, 1 KIT
CAS nummer:  
Price
2.720,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
The His·Bind buffer kit is a set of pre-tested buffers designed for use with IDA-based His·Bind resin for convenient, rapid one-step purification of proteins by metal chelation chromatography.
Solutions are included for Ni²⁺ charging, binding, washing and elution of up to ten 2,5 ml columns.
Leveringsinformation: The kit contains 8× binding buffer (2×80 ml), 8× wash buffer (25 ml), 4× elute buffer (50 ml), 4× strip buffer (50 ml) and 8× charge buffer (20 ml).
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    His·Bind® buffer kit