Fittings, InfinityLab
InfinityLab quick connect LC fitting, Rustfrit stål, —
Katalog # 558-0637
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
InfinityLab Quick Connect fittings are 'finger-tight' reusable fittings for UHPLC, stable to 1300 bar.
- No tools needed
For harder to reach areas on your LC instrument, the InfinityLab Quick Turn fitting produces either a finger-tight connection (stable to 600 bar), or a premium UHPLC connection (stable to 1300 bar), with a quick turn of the wrench.
Specifikationer
- Beskrivelse:InfinityLab quick connect LC fitting
- Længde:—
- Materiale:Rustfrit stål
- Seq:14
- Cat. No.:558-0637