Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2,4'-DDT

2,4'-DDT

2,4'-DDT, 1000 µg/ml in methanol, 1 ml/ampoule, 1
Katalog # REST32200
Leverandør:   Restek
2,4'-DDT
Katalog # REST32200
Leverandør:  Restek
CAS nummer:  
2,4'-DDT
2,4'-DDT

2,4'-DDT, 1000 µg/ml in methanol, 1 ml/ampoule, 1

CAS nummer:  
Price
482,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifikationer

Specifikationer

  • Koncentration:
    1000 µg/ml in methanol
  • Pakn:
    1
  • Beskrivelse:
    2,4'-DDT
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST32200
  • CAS-nr:
    789-02-6
  • Formel:
    C₁₄H₉Cl₅

Specifikationer