Orders LinkContactUsLinkComponent
PCB congener standard #2 (7 components)
PCB congener standard #2 (7 components)
PCB congener standard, 10 µg/ml each in isooctane, 1 ml/ampoule, 1 mL
Katalog # REST32294
Leverandør:   Restek
PCB congener standard #2 (7 components)
Katalog # REST32294
Leverandør:  Restek
CAS nummer:  
PCB congener standard #2 (7 components)
PCB congener standard #2 (7 components)
PCB congener standard, 10 µg/ml each in isooctane, 1 ml/ampoule, 1 mL
CAS nummer:  
Price
605,00 kr
Log ind for tilgængelighed
1 * 1 mL
Product Details & Documents
Contains: 2,4,4'-trichlorobiphenyl (BZ #28), 2,2',5,5'-tetrachlorobiphenyl (BZ #52), 2,2',4,5,5'-pentachlorobiphenyl (BZ #101), 2,3',4,4',5-pentachlorobiphenyl (BZ #118), 2,2',3,4,4',5'-hexachlorobiphenyl (BZ #138), 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153), 2,2',3,4,4',5,5'-heptachlorobiphenyl (BZ #180).
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Specifikationer
Specifikationer
  • Koncentration:
    10 µg/ml each in isooctane
  • Pakn:
    1 mL
  • Beskrivelse:
    PCB congener standard
  • Pk Info:
    1 ml/ampoule
Specifikationer