Zero dilution inner liner
Katalog # REST22991
Supplier: Restek
Specifications
- Udv.Ø:2,0 mm
- Ø indv.:1,0 mm
- Beskrivelse:Zero dilution inner liner
- Længde:73 mm
- Pakn:5
Specifications
About this item
This liner is ideal for PerkinElmer auto SYS and clarus 580/680 GCs equipped with split/splitless inlets.
Made from borosilicate glass.