FAME #9 mix
FAME #9, each mixture has minimum 50 mg/ampoule, Ampoule, 1
REST35018
: Restek
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
- Suitable for oils of mid-range to long chain lengths (C16 - C20)
This mix contains five components such as C16:00 methyl palmitate (20%), C17:00 methyl heptadecanoate (20%), C18:00 methyl stearate (20%), C19:00 methyl nonadecanoate (20%), C20:00 methyl arachidate (20%).
- Pakning:Ampoule
- Pakn:1
- Beskrivelse:FAME #9, each mixture has minimum 50 mg/ampoule