Orders LinkContactUsLinkComponent
FAME #5 mix
FAME #5 mix
FAME #5, each mixture has minimum 50 mg/ampoule, Ampoule, 1
Katalog # REST35014
Leverandør:   Restek
FAME #5 mix
Katalog # REST35014
Leverandør:  Restek
CAS nummer:  
FAME #5 mix
FAME #5 mix
FAME #5, each mixture has minimum 50 mg/ampoule, Ampoule, 1
CAS nummer:  
Price
683,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for oils of mid-range to long chain lengths (C16 - C24)
This mix contains five components such as C16:01 methyl palmitoleate (20%), C18:01 methyl oleate (20%), C20:01 methyl eicosenoate (20%), C22:01 methyl erucate (20%), C24:01:00 methyl nervonate (20%).
Specifikationer
Specifikationer
  • Pakning:
    Ampoule
  • Pakn:
    1
  • Beskrivelse:
    FAME #5, each mixture has minimum 50 mg/ampoule