Orders LinkContactUsLinkComponent
FAME #2 mix
FAME #2 mix
FAME #2, each mixture has minimum 50 mg/ampoule, Ampoule, 1
Katalog # REST35011
Leverandør:   Restek
FAME #2 mix
Katalog # REST35011
Leverandør:  Restek
CAS nummer:  
FAME #2 mix
FAME #2 mix
FAME #2, each mixture has minimum 50 mg/ampoule, Ampoule, 1
CAS nummer:  
Price
698,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for oils of short to mid-range chain lengths (C6 - C14)
This mix contains five components such as C6:00 methyl caproate (20%), C8:00 methyl caprylate (20%), C10:00 methyl caprate (20%), C12:00 methyl laurate (20%), C14:00 methyl myristate (20%).
Specifikationer
Specifikationer
  • Pakning:
    Ampoule
  • Pakn:
    1
  • Beskrivelse:
    FAME #2, each mixture has minimum 50 mg/ampoule